accessibility menu, dialog, popup

UserName

HClClNHN

CAS RN 303-26-4

IUPAC Name: hydrogen 1-[(4-chlorophenyl)(phenyl)methyl]piperazine chloride
Synonyms: hydrogen 1-[(4-chlorophenyl)(phenyl)methyl]piperazine chloride
Molecular Weight (g/mol): 323.26
Molecular Formula: C17H20Cl2N2
InChi Key: XFIUZNUONMJUIH-UHFFFAOYNA-N
SMILES: [H+].[Cl-].ClC1=CC=C(C=C1)C(N1CCNCC1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (5)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)