accessibility menu, dialog, popup

UserName

CAS RN 30414-53-0

H3COOOCH3

CAS RN 30414-53-0

IUPAC Name: methyl 3-oxopentanoate
Synonyms: methyl 3-oxopentanoate
Molecular Weight (g/mol): 130.14
Molecular Formula: C6H10O3
InChi Key: XJMIXEAZMCTAGH-UHFFFAOYSA-N
SMILES: CCC(=O)CC(=O)OC
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)