CAS RN 307002-73-9
CAS RN 307002-73-9
IUPAC Name:
N-[2-(4-{benzyl[2-(hydroxycarbamoyl)-4,6-dimethylphenyl]sulfamoyl}phenoxy)ethyl]-1-benzofuran-2-carboxamide
Synonyms:
N-[2-(4-{benzyl[2-(hydroxycarbamoyl)-4,6-dimethylphenyl]sulfamoyl}phenoxy)ethyl]-1-benzofuran-2-carboxamide
Molecular Weight (g/mol):
613.69
Molecular Formula:
C33H31N3O7S
InChi Key:
FARMEEAGJWMFSZ-UHFFFAOYSA-N
SMILES:
CC1=CC(C)=C(N(CC2=CC=CC=C2)S(=O)(=O)C2=CC=C(OCCNC(=O)C3=CC4=CC=CC=C4O3)C=C2)C(=C1)C(=O)NO