CAS RN 30796-85-1
CAS RN 30796-85-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-(5-Methyl-2-furyl)ethanamine, TRC
CAS: 30796-85-1 Molecular Formula: C7H11NO Molecular Weight (g/mol): 125.16 Synonym: 5-Methyl-2-furanethanamine,2-(5-Methylfuran-2-yl)ethan-1-amine,2-(5-Methylfuran-2-yl)ethanamine,2-(5-Methylfuran-2-yl)ethylamine,beta-(5-Methyl-2-furanyl)ethanamine IUPAC Name: 2-(5-methylfuran-2-yl)ethanamine SMILES: Cc1oc(CCN)cc1