accessibility menu, dialog, popup

UserName

CAS RN 3107-19-5

ONOClFCl

CAS RN 3107-19-5

IUPAC Name: 1,3-dichloro-2-fluoro-5-nitrobenzene
Synonyms: 1,3-dichloro-2-fluoro-5-nitrobenzene
Molecular Weight (g/mol): 209.99
Molecular Formula: C6H2Cl2FNO2
InChi Key: VMAATSFMXSMKPG-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Cl)=C(F)C(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results