accessibility menu, dialog, popup

UserName

CAS RN 3133-81-1

H3COOSH3C

CAS RN 3133-81-1

IUPAC Name: methyl 3-methyl-1-benzothiophene-2-carboxylate
Synonyms: methyl 3-methyl-1-benzothiophene-2-carboxylate
Molecular Weight (g/mol): 206.26
Molecular Formula: C11H10O2S
InChi Key: FDBXIMKCWBPTDG-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C)C2=CC=CC=C2S1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results