CAS RN 315706-13-9
CAS RN 315706-13-9
IUPAC Name:
2-{2-[4-(3,4-dichlorophenyl)-1,3-thiazol-2-yl]hydrazin-1-ylidene}-3-(2-nitrophenyl)propanoic acid
Synonyms:
2-{2-[4-(3,4-dichlorophenyl)-1,3-thiazol-2-yl]hydrazin-1-ylidene}-3-(2-nitrophenyl)propanoic acid
Molecular Weight (g/mol):
451.28
Molecular Formula:
C18H12Cl2N4O4S
InChi Key:
KFRKRECSIYXARE-UHFFFAOYSA-N
SMILES:
OC(=O)C(CC1=CC=CC=C1[N+]([O-])=O)=NNC1=NC(=CS1)C1=CC(Cl)=C(Cl)C=C1
1
–
2
of
2
results
1
Tocris Bioscience™ 4EGI-1
CAS: 315706-13-9 Molecular Formula: C18H12Cl2N4O4S
2
4EGI-1, MedChemExpress
MedChemExpress 4EGI-1 is an inhibitor of eIF4E/eIF4G interaction, with a Kd of 25 μM against eIF4E binding.