CAS RN 31842-01-0
CAS RN 31842-01-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Indoprofen, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
2
Indoprofen, MedChemExpress
MedChemExpress Indoprofen is a non-steroidal anti-inflammatory drug, provide insight into treatments for spinal muscular atrophies.