accessibility menu, dialog, popup

UserName

CAS RN 32024-15-0

H3COOIOCH3

CAS RN 32024-15-0

IUPAC Name: 3-iodo-4,5-dimethoxybenzaldehyde
Synonyms: 3-iodo-4,5-dimethoxybenzaldehyde
Molecular Weight (g/mol): 292.07
Molecular Formula: C9H9IO3
InChi Key: MVPNBXPAUYYZAF-UHFFFAOYSA-N
SMILES: COC1=C(OC)C(I)=CC(C=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results