CAS RN 32113-85-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Methyl Acetate-d6, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
2
Methyl Acetate-d6, TRC
CAS: 32113-85-2 Molecular Formula: C3 D6 O2 Molecular Weight (g/mol): 80.12 Synonym: Acetic-d3 acid, methyl-d3 ester (6CI, 8CI, 9CI),Methyl Acetate-d6,Methyl Acetate-D6,Acetic-d3 acid methyl-d3 ester,Acetic acid methyl ester-d6 IUPAC Name: trideuteriomethyl SMILES: [2H]C([2H])([2H])OC(=O)C([2H])([2H])[2H]