accessibility menu, dialog, popup

UserName

CAS RN 3222-50-2

CH3NHOO

CAS RN 3222-50-2

IUPAC Name: 4-methylpyridine-3-carboxylic acid
Synonyms: 4-methylpyridine-3-carboxylic acid
Molecular Weight (g/mol): 137.14
Molecular Formula: C7H7NO2
InChi Key: ZKUZSTXNVMIDCY-UHFFFAOYSA-N
SMILES: CC1=CC=NC=C1C(O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results