accessibility menu, dialog, popup

UserName

CAS RN 3228-99-7

ClClClCl

CAS RN 3228-99-7

IUPAC Name: 1,3-dichloro-2,2-bis(chloromethyl)propane
Synonyms: tetrachloride (tetrachloromethyl)methane
Molecular Weight (g/mol): 209.92
Molecular Formula: C5H8Cl4
InChi Key: KPZGRMZPZLOPBS-UHFFFAOYSA-N
SMILES: ClCC(CCl)(CCl)CCl
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)