CAS RN 326-64-7
IUPAC Name:
5-chloro-2-(trifluoromethoxy)aniline
Synonyms:
5-chloro-2-(trifluoromethoxy)aniline
Molecular Weight (g/mol):
211.57
Molecular Formula:
C7H5ClF3NO
InChi Key:
KSBUTBXMRFVLND-UHFFFAOYSA-N
SMILES:
NC1=C(OC(F)(F)F)C=CC(Cl)=C1
1
–
2
of
2
results
2
5-Chloro-2-(trifluoromethoxy)aniline, 97%, Thermo Scientific™
CAS: 326-64-7 Molecular Formula: C7H5ClF3NO Molecular Weight (g/mol): 211.568 MDL Number: MFCD06410919 InChI Key: KSBUTBXMRFVLND-UHFFFAOYSA-N Synonym: 5-chloro-2-trifluoromethoxy aniline,5-chloro-2-trifluoro methoxy aniline,5-chloro-2-trifluoromethoxy benzenamine,benzenamine, 5-chloro-2-trifluoromethoxy,5-chloro-2-trifluoromethoxy-phenylamine,2-amino-4-chlorotrifluoro-methoxybenzene,5-chloro-2-trifluoromethoxy phenylamine PubChem CID: 10932748 IUPAC Name: 5-chloro-2-(trifluoromethoxy)aniline SMILES: C1=CC(=C(C=C1Cl)N)OC(F)(F)F