accessibility menu, dialog, popup

UserName

CAS RN 32858-93-8

OHOFFF

CAS RN 32858-93-8

IUPAC Name: 2-(trifluoromethoxy)phenol
Synonyms: 2-(trifluoromethoxy)phenol
Molecular Weight (g/mol): 178.11
Molecular Formula: C7H5F3O2
InChi Key: GQWMNVOVQZIPJC-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1OC(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results