accessibility menu, dialog, popup

UserName

CH3ClHOOF

CAS RN 32890-90-7

IUPAC Name: 6-chloro-2-fluoro-3-methylbenzoic acid
Synonyms: 6-chloro-2-fluoro-3-methylbenzoic acid
Molecular Weight (g/mol): 188.58
Molecular Formula: C8H6ClFO2
InChi Key: HLQCBMOWEQKVBL-UHFFFAOYSA-N
SMILES: CC1=CC=C(Cl)C(C(O)=O)=C1F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results