accessibility menu, dialog, popup

UserName

CH3OOFNOO

CAS RN 329-59-9

IUPAC Name: methyl 4-fluoro-3-nitrobenzoate
Synonyms: methyl 4-fluoro-3-nitrobenzoate
Molecular Weight (g/mol): 199.14
Molecular Formula: C8H6FNO4
InChi Key: CNJJSTPBUHAEFH-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=C(F)C=C1)[N+]([O-])=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results