accessibility menu, dialog, popup

UserName

H2NHN

CAS RN 3312-60-5

IUPAC Name: N1-cyclohexylpropane-1,3-diamine
Synonyms: N1-cyclohexylpropane-1,3-diamine
Molecular Weight (g/mol): 156.27
Molecular Formula: C9H20N2
InChi Key: ITZPOSYADVYECJ-UHFFFAOYSA-N
SMILES: NCCCNC1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results