accessibility menu, dialog, popup

UserName

ClNClCl

CAS RN 33216-52-3

IUPAC Name: 3,4,5-trichloropyridine
Synonyms: 3,4,5-trichloropyridine
Molecular Weight (g/mol): 182.43
Molecular Formula: C5H2Cl3N
InChi Key: KKWRVUBDCJQHBZ-UHFFFAOYSA-N
SMILES: ClC1=CN=CC(Cl)=C1Cl
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)