accessibility menu, dialog, popup

UserName

CAS RN 33224-23-6

H3CONNOO

CAS RN 33224-23-6

IUPAC Name: 4-methoxy-3-nitrobenzonitrile
Synonyms: 4-methoxy-3-nitrobenzonitrile
Molecular Weight (g/mol): 178.15
Molecular Formula: C8H6N2O3
InChi Key: ACAYRJJVKPMSIS-UHFFFAOYSA-N
SMILES: COC1=C(C=C(C=C1)C#N)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results