CAS RN 33240-34-5
CAS RN 33240-34-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
173.34
Molecular Formula:
C5H9BrMg
InChi Key:
JMPXNLAYOWCGCW-UHFFFAOYSA-M
SMILES:
[Mg++].[Br-].C1CC[CH-]C1
1
–
1
of
1
results
1
Promotions Available
Cyclopentylmagnesium bromide, 2.0M solution in diethyl ether, AcroSeal™
Cyclopentylmagnesium bromide, 2.0M solution in diethyl ether, AcroSeal™
CAS: 33240-34-5 Molecular Formula: C5H9BrMg Molecular Weight (g/mol): 173.34 MDL Number: MFCD00209569 InChI Key: PZOJUJCALDFFHC-UHFFFAOYSA-M Synonym: cyclopentylmagnesium bromide,bromocyclopentylmagnesium,magnesium, bromocyclopentyl,cyclopentyl magnesium bromide,bromo cyclopentyl magnesium,cyclopentylmagnesium bromide solution,cydopentylmagnesium bromide,cyclopentylmagnsium bromide,pzojujcaldffhc-uhfffaoysa-m,cyclopentylmagnesium bromide solution, 2.0 m in diethyl ether,grignard reagent PubChem CID: 118418 IUPAC Name: magnesium;cyclopentane;bromide SMILES: Br[Mg]C1CCCC1