accessibility menu, dialog, popup

UserName

CAS RN 333-49-3

NH2NSHN

CAS RN 333-49-3

IUPAC Name: 6-amino-1,2-dihydropyrimidine-2-thione
Synonyms: 4-amino-3H-pyrimidine-2-thione
Molecular Weight (g/mol): 127.17
Molecular Formula: C4H5N3S
InChi Key: DCPSTSVLRXOYGS-UHFFFAOYSA-N
SMILES: NC1=CC=NC(=S)N1
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results