accessibility menu, dialog, popup

UserName

CAS RN 33332-28-4

NH2NClN

CAS RN 33332-28-4

IUPAC Name: 6-chloropyrazin-2-amine
Synonyms: 2-amino-6-chloropyrazine C4H4ClN3
Molecular Weight (g/mol): 129.55
Molecular Formula: C4H4ClN3
InChi Key: JTPXVCKCLBROOJ-UHFFFAOYSA-N
SMILES: NC1=CN=CC(Cl)=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results