CAS RN 333353-44-9
CAS RN 333353-44-9
IUPAC Name:
N'-(4-chlorophenyl)-N-(2,2,6,6-tetramethylpiperidin-4-yl)ethanediamide
Synonyms:
N'-(4-chlorophenyl)-N-(2,2,6,6-tetramethylpiperidin-4-yl)ethanediamide
Molecular Weight (g/mol):
337.85
Molecular Formula:
C17H24ClN3O2
InChi Key:
ZKXLQCIOURANAD-UHFFFAOYSA-N
SMILES:
CC1(C)CC(CC(C)(C)N1)NC(=O)C(=O)NC1=CC=C(Cl)C=C1
1
–
1
of
1
results
1
NBD-556, MedChemExpress
MedChemExpress NBD-556, a CD4 mimetic, is a potent HIV-1 entry inhibitor that blocks the gp120-CD4 interaction. NBD-556 shows potent cell fusion and virus-cell fusion inhibitory activity at low micromolar levels.