CAS RN 335-56-8
IUPAC Name:
1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexane
Synonyms:
1-bromo-tridecafluorohexane
Molecular Weight (g/mol):
398.95
Molecular Formula:
C6BrF13
InChi Key:
JTYRBFORUCBNHJ-UHFFFAOYSA-N
SMILES:
FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)Br
1
–
2
of
2
results
1
Perfluorohexyl bromide, 97%
CAS: 335-56-8 Molecular Formula: C6BrF13 Molecular Weight (g/mol): 398.95 InChI Key: JTYRBFORUCBNHJ-UHFFFAOYSA-N Synonym: 1-bromoperfluorohexane,perfluorohexyl bromide,perfluorohexylbromide,tridecafluorohexyl bromide,1-bromo-tridecafluorohexane,bromoperfluorohexane,acmc-1cmhf,1-bromotridecafluorohexane,tridecafluoro-1-bromohexane PubChem CID: 92755 IUPAC Name: 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexane SMILES: C(C(C(C(F)(F)Br)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F