accessibility menu, dialog, popup

UserName

H3CNOCH3HOHCl

CAS RN 33625-43-3

IUPAC Name: hydrogen 2,5-dimethyl-4-[(morpholin-4-yl)methyl]phenol chloride
Synonyms: hydrogen 2,5-dimethyl-4-(morpholin-4-ylmethyl)phenol chloride
Molecular Weight (g/mol): 257.76
Molecular Formula: C13H20ClNO2
InChi Key: RLGUYDSHQWRANT-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1=CC(CN2CCOCC2)=C(C)C=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results