CAS RN 338-03-4
CAS RN 338-03-4
IUPAC Name:
(3S)-4,4,4-trifluoro-3-hydroxy-3-methylbutanoate
Synonyms:
(3S)-4,4,4-trifluoro-3-hydroxy-3-methylbutanoate
Molecular Weight (g/mol):
171.1
Molecular Formula:
C5H6F3O3
InChi Key:
JCJGOWYCFRASTM-BYPYZUCNSA-M
SMILES:
C[C@](O)(CC([O-])=O)C(F)(F)F
1
–
1
of
1
results
1
4,4,4-Trifluoro-3-hydroxy-3-methylbutyric acid, 98%
CAS: 338-03-4 Molecular Formula: C5H7F3O3 Molecular Weight (g/mol): 172.103 MDL Number: MFCD00039531 InChI Key: JCJGOWYCFRASTM-UHFFFAOYSA-N Synonym: 3-trifluoromethyl-3-hydroxybutyric acid,4,4,4-trifluoro-3-hydroxy-3-methylbutyric acid,3-hydroxy-3-methyl-4,4,4-trifluorobutyric acid,4,4,4-trifluoro-3-hydroxy-3-methyl-butyric acid,butanoic acid, 4,4,4-trifluoro-3-hydroxy-3-methyl,pubchem12644,acmc-20an2r,3-hydroxy-3-trifluoromethylbutyric acid,3-hydroxy-3-trifluoromethyl butyric acid PubChem CID: 136168 IUPAC Name: 4,4,4-trifluoro-3-hydroxy-3-methylbutanoic acid SMILES: CC(CC(=O)O)(C(F)(F)F)O