accessibility menu, dialog, popup

UserName

CAS RN 33863-76-2

ClBrF

CAS RN 33863-76-2

IUPAC Name: 1-bromo-3-chloro-5-fluorobenzene
Synonyms: 1-bromo-3-chloro-5-fluorobenzene
Molecular Weight (g/mol): 209.44
Molecular Formula: C6H3BrClF
InChi Key: GGMDFPMASIXEIR-UHFFFAOYSA-N
SMILES: FC1=CC(Cl)=CC(Br)=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)