accessibility menu, dialog, popup

UserName

H3COClO

CAS RN 33863-86-4

IUPAC Name: 4-butoxybenzoyl chloride
Synonyms: 4-butoxybenzoyl chloride
Molecular Weight (g/mol): 212.67
Molecular Formula: C11H13ClO2
InChi Key: KMGCTFHTBKBITO-UHFFFAOYSA-N
SMILES: CCCCOC1=CC=C(C=C1)C(Cl)=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)