accessibility menu, dialog, popup

UserName

NH2HNOFFF

CAS RN 339-59-3

IUPAC Name: 4-(trifluoromethyl)benzohydrazide
Synonyms: 4-(trifluoromethyl)benzohydrazide
Molecular Weight (g/mol): 204.15
Molecular Formula: C8H7F3N2O
InChi Key: GKBDXTNCBPZMFX-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=C(C=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results