accessibility menu, dialog, popup

UserName

ON

CAS RN 33985-71-6

IUPAC Name: 1-azatricyclo[7.3.1.0⁵,¹³]trideca-5,7,9(13)-triene-7-carbaldehyde
Synonyms: 1-azatricyclo[7.3.1.0⁵,¹³]trideca-5,7,9(13)-triene-7-carbaldehyde
Molecular Weight (g/mol): 201.27
Molecular Formula: C13H15NO
InChi Key: XIIVBURSIWWDEO-UHFFFAOYSA-N
SMILES: O=CC1=CC2=C3N(CCCC3=C1)CCC2
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)