CAS RN 34042-85-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Sudoxicam, TRC
CAS: 34042-85-8 Molecular Formula: C13 H11 N3 O4 S2 Molecular Weight (g/mol): 337.37 Synonym: 4-Hydroxy-2-methyl-N-2-thiazolyl-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide; IUPAC Name: 4-hydroxy-2-methyl-1,1-dioxo-N-(1,3-thiazol-2-yl)-1λ^{6},2-benzothiazine-3-carboxamide SMILES: CN1C(=C(O)c2ccccc2S1(=O)=O)C(=O)Nc3nccs3