accessibility menu, dialog, popup

UserName

CAS RN 34064-86-3

NNHNH2

CAS RN 34064-86-3

IUPAC Name: 4-(2-cyanoethyl)piperazine-1,4-diium
Synonyms: 4-(2-cyanoethyl)piperazine-1,4-diium
Molecular Weight (g/mol): 141.22
Molecular Formula: C7H15N3
InChi Key: MVOFPBMQTXKONX-UHFFFAOYSA-P
SMILES: N#CCC[NH+]1CC[NH2+]CC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results