accessibility menu, dialog, popup

UserName

CAS RN 34118-92-8

HClH3CNCH3NHOHNCH3O

CAS RN 34118-92-8

IUPAC Name: hydrogen N-[2-(diethylamino)ethyl]-4-acetamidobenzamide chloride
Synonyms: N-acetylprocainamide
Molecular Weight (g/mol): 313.83
Molecular Formula: C15H24ClN3O2
InChi Key: IYEWBJUCJHKLHD-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCN(CC)CCNC(=O)C1=CC=C(NC(C)=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)