CAS RN 34124-14-6
IUPAC Name:
2-[2-(2-aminophenyl)ethyl]aniline
Synonyms:
2-[2-(2-aminophenyl)ethyl]aniline
Molecular Weight (g/mol):
212.3
Molecular Formula:
C14H16N2
InChi Key:
ZYHQGITXIJDDKC-UHFFFAOYSA-N
SMILES:
NC1=CC=CC=C1CCC1=CC=CC=C1N
1
–
1
of
1
results
1
2,2'-Ethylenedianiline, TRC
CAS: 34124-14-6 Molecular Formula: C14H16N2 Molecular Weight (g/mol): 212.29 Synonym: 2,2'-(1,2-Ethanediyl)bisbenzenamine,alpha,alpha'-Bi-o-toluidine,2,2'-Diaminobibenzyl,2,2'-Diaminodibenzyl,NSC 159119 SMILES: Nc1ccccc1CCc2ccccc2N