accessibility menu, dialog, popup

UserName

NBrOH(E/Z)

CAS RN 34158-72-0

IUPAC Name: (Z)-N-[(2-bromophenyl)methylidene]hydroxylamine
Synonyms: (Z)-N-[(2-bromophenyl)methylidene]hydroxylamine
Molecular Weight (g/mol): 200.04
Molecular Formula: C7H6BrNO
InChi Key: PSIRFUPZHPEKAE-UITAMQMPSA-N
SMILES: O\N=C/C1=CC=CC=C1Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results