accessibility menu, dialog, popup

UserName

CAS RN 3430-21-5

CH3BrHNH2N

CAS RN 3430-21-5

IUPAC Name: 2-amino-5-bromo-3-methylpyridin-1-ium
Synonyms: 2-amino-5-bromo-3-methylpyridin-1-ium
Molecular Weight (g/mol): 188.05
Molecular Formula: C6H8BrN2
InChi Key: KBLGGRWUEVCNPY-UHFFFAOYSA-O
SMILES: CC1=CC(Br)=C[NH+]=C1N
Quantity 
  • (2)
  • (3)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)