accessibility menu, dialog, popup

UserName

OHNNOO(Z)

CAS RN 3431-62-7

IUPAC Name: (Z)-N-[(3-nitrophenyl)methylidene]hydroxylamine
Synonyms: (Z)-N-[(3-nitrophenyl)methylidene]hydroxylamine
Molecular Weight (g/mol): 166.14
Molecular Formula: C7H6N2O3
InChi Key: GQMMRLBWXCGBEV-YVMONPNESA-N
SMILES: O\N=C/C1=CC=CC(=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results