CAS RN 344-38-7
IUPAC Name:
4-bromo-1-nitro-2-(trifluoromethyl)benzene
Synonyms:
4-bromo-1-nitro-2-(trifluoromethyl)benzene
Molecular Weight (g/mol):
270.01
Molecular Formula:
C7H3BrF3NO2
InChi Key:
ZHLYHEDQTJZYFI-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=C(Br)C=C1C(F)(F)F
1
–
2
of
2
results
1
5-Bromo-2-nitrobenzotrifluoride, 98+%
CAS: 344-38-7 Molecular Formula: C7H3BrF3NO2 Molecular Weight (g/mol): 270.005 MDL Number: MFCD00014706 InChI Key: ZHLYHEDQTJZYFI-UHFFFAOYSA-N Synonym: 5-bromo-2-nitrobenzotrifluoride,4-bromo-1-nitro-2-trifluoromethyl benzene,2-nitro-5-bromobenzotrifluoride,5-bromo-2-nitro-a,a,a-trifluorotoluene,5-bromo-a-a,a-trifluoro-2-nitrotoluene,4-bromo-1-nitro-2 trifluoromethyl benzene,4-bromo-1-nitro-2-trifluoromethyl-benzene,benzene, 4-bromo-1-nitro-2-trifluoromethyl,5-bromo-alpha,alpha,alpha-trifluoro-2-nitrotoluene,2-nitro-5-bromotrifluorotoluol PubChem CID: 136171 IUPAC Name: 4-bromo-1-nitro-2-(trifluoromethyl)benzene SMILES: C1=CC(=C(C=C1Br)C(F)(F)F)[N+](=O)[O-]