CAS RN 344591-91-9
CAS RN 344591-91-9
IUPAC Name:
[1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]boronic acid
Synonyms:
2-methyl-5-(trifluoromethyl)pyrazol-3-ylboronic acid
Molecular Weight (g/mol):
193.92
Molecular Formula:
C5H6BF3N2O2
InChi Key:
XPUNXPPXMANQGF-UHFFFAOYSA-N
SMILES:
CN1N=C(C=C1B(O)O)C(F)(F)F