accessibility menu, dialog, popup

UserName

H3CCH3NH2

CAS RN 34566-04-6

IUPAC Name: octan-2-amine
Synonyms: 2-octanamine
Molecular Weight (g/mol): 129.25
Molecular Formula: C8H19N
InChi Key: HBXNJMZWGSCKPW-UHFFFAOYNA-N
SMILES: CCCCCCC(C)N
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results