accessibility menu, dialog, popup

UserName

(R)ONH2OHHSHCl

CAS RN 345909-32-2

IUPAC Name: (2R)-2-azaniumyl-3-sulfanylpropanoate
Synonyms: L-cysteine zwitterion
Molecular Weight (g/mol): 121.15
Molecular Formula: C3H7NO2S
InChi Key: XUJNEKJLAYXESH-REOHCLBHSA-N
SMILES: [NH3+][C@@H](CS)C([O-])=O
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)