accessibility menu, dialog, popup

UserName

CAS RN 34592-47-7

OO(R)SH2N

CAS RN 34592-47-7

IUPAC Name: (4R)-1,3-thiazolidin-3-ium-4-carboxylate
Synonyms: (4R)-1,3-thiazolidin-3-ium-4-carboxylate
Molecular Weight (g/mol): 133.17
Molecular Formula: C4H7NO2S
InChi Key: DZLNHFMRPBPULJ-VKHMYHEASA-N
SMILES: [O-]C(=O)[C@@H]1CSC[NH2+]1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)