accessibility menu, dialog, popup

UserName

BrNN

CAS RN 34658-66-7

IUPAC Name: 2-(4-bromophenyl)imidazo[1,2-a]pyridine
Synonyms: 2-(4-bromophenyl)imidazo[1,2-a]pyridine
Molecular Weight (g/mol): 273.13
Molecular Formula: C13H9BrN2
InChi Key: GRZUOGFRIHABDK-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C1=CN2C=CC=CC2=N1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results