accessibility menu, dialog, popup

UserName

CAS RN 34859-65-9

H3COOSNH3CNH2

CAS RN 34859-65-9

IUPAC Name: ethyl 5-amino-3-methyl-1,2-thiazole-4-carboxylate
Synonyms: ethyl 5-amino-3-methyl-1,2-thiazole-4-carboxylate
Molecular Weight (g/mol): 186.23
Molecular Formula: C7H10N2O2S
InChi Key: ZDGBWOZFPKNORL-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)SN=C1C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results