accessibility menu, dialog, popup

UserName

CAS RN 350-57-2

FFFFO

CAS RN 350-57-2

IUPAC Name: (1,1,2,2-tetrafluoroethoxy)benzene
Synonyms: (1,1,2,2-tetrafluoroethoxy)benzene
Molecular Weight (g/mol): 194.13
Molecular Formula: C8H6F4O
InChi Key: GRDIVJPQARIBNZ-UHFFFAOYSA-N
SMILES: FC(F)C(F)(F)OC1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results