CAS RN 3505-38-2
CAS RN 3505-38-2
IUPAC Name:
(2Z)-but-2-enedioic acid; {2-[(4-chlorophenyl)(pyridin-2-yl)methoxy]ethyl}dimethylamine
Synonyms:
p-carbinoxamine maleate paracarbinoxamine maleate
Molecular Weight (g/mol):
406.86
Molecular Formula:
C20H23ClN2O5
InChi Key:
GVNWHCVWDRNXAZ-BTJKTKAUNA-N
SMILES:
OC(=O)\C=C/C(O)=O.CN(C)CCOC(C1=CC=C(Cl)C=C1)C1=CC=CC=N1
1
–
3
of
3
results
1
Carbinoxamine Maleate, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
2
Carbinoxamine Maleate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
Carbinoxamine maleate salt, MedChemExpress
MedChemExpress Carbinoxamine maleate salt is a histamine H1 receptor antagonist.