accessibility menu, dialog, popup

UserName

CAS RN 351-87-1

H3COOHNFFF

CAS RN 351-87-1

IUPAC Name: 3-oxo-N-[4-(trifluoromethyl)phenyl]butanamide
Synonyms: 3-oxo-N-[4-(trifluoromethyl)phenyl]butanamide
Molecular Weight (g/mol): 245.2
Molecular Formula: C11H10F3NO2
InChi Key: GGDAWUSJZZHVBR-UHFFFAOYSA-N
SMILES: CC(=O)CC(=O)NC1=CC=C(C=C1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)