accessibility menu, dialog, popup

UserName

SOOClFBr

CAS RN 351003-51-5

IUPAC Name: 4-bromo-3-fluorobenzene-1-sulfonyl chloride
Synonyms: 4-bromo-3-fluorobenzenesulfonyl chloride
Molecular Weight (g/mol): 273.5
Molecular Formula: C6H3BrClFO2S
InChi Key: IZCXVIACMHTZNA-UHFFFAOYSA-N
SMILES: FC1=C(Br)C=CC(=C1)S(Cl)(=O)=O
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)