accessibility menu, dialog, popup

UserName

CAS RN 35141-30-1

H3COSiHNNHNH2OCH3OCH3

CAS RN 35141-30-1

IUPAC Name: 3,3-dimethoxy-2-oxa-7,10-diaza-3-siladodecan-12-amine
Synonyms: 3,3-dimethoxy-2-oxa-7,10-diaza-3-siladodecan-12-amine
Molecular Weight (g/mol): 265.43
Molecular Formula: C10H27N3O3Si
InChi Key: NHBRUUFBSBSTHM-UHFFFAOYSA-N
SMILES: CO[Si](CCCNCCNCCN)(OC)OC
Boiling Point 
  • (3)
Grade 
  • (3)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)