accessibility menu, dialog, popup

UserName

H3COOSH2N

CAS RN 35212-85-2

IUPAC Name: methyl 3-amino-1-benzothiophene-2-carboxylate
Synonyms: methyl 3-amino-1-benzothiophene-2-carboxylate
Molecular Weight (g/mol): 207.25
Molecular Formula: C10H9NO2S
InChi Key: VLHHEYMZLXKSQO-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(N)C2=CC=CC=C2S1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)